siRNA Details
Gene Symbol: Olfr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011662.2 | NP_001011662.2 | 2 | 677 |
Gene Symbol: Olfr178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146997.2 | NP_667208.2 | 1 | 495 | ||
GenBank mRNA | BC111099.1 | - | 1 | 494 | ||
BC127221.1 | - | 1 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
