siRNA Details
Gene Symbol: Olfr406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011863.1 | NP_001011863.1 | 1 | 766 | ||
GenBank mRNA | BC147149.1 | - | 1 | 813 | 9056162 | |
BC147150.1 | - | 1 | 813 | 9056170 |
Gene Symbol: Olfr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011002.2 | NP_035132.2 | 2 | 1095 | ||
GenBank mRNA | AF102538.1 | - | 1 | 577 | ||
BC139159.1 | - | 1 | 855 | 8862181 | ||
BC139161.1 | - | 1 | 855 | 8862183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
