siRNA Details
Gene Symbol: Olfr100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207673.1 | NP_997556.1 | 1 | 254 | ||
GenBank mRNA | BC111097.1 | - | 1 | 253 | ||
BC127233.1 | - | 1 | 254 | 40054913 | ||
BC127234.1 | - | 1 | 254 | 40054914 |
