siRNA Details
Gene Symbol: Olfr310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011520.2 | NP_001011520.2 | 1 | 97 | ||
GenBank mRNA | BC147445.1 | - | 1 | 97 | 9056087 | |
BC147455.1 | - | 1 | 97 | 9056097 |
