siRNA Details
Gene Symbol: Olfr645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207144.1 | NP_997027.1 | 1 | 464 | ||
GenBank mRNA | BC132009.1 | - | 1 | 562 | 40130286 | |
BC132011.1 | - | 1 | 563 | 40130288 |
