siRNA Details
Gene Symbol: Olfr576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001805.2 | NP_001001805.2 | 1 | 480 | ||
GenBank mRNA | BC106977.2 | - | 1 | 480 | 40042102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
