siRNA Details
Gene Symbol: Olfr1313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207150.1 | NP_997033.1 | 1 | 125 | ||
GenBank mRNA | BC127964.1 | - | 1 | 129 | 40134976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
