siRNA Details
Gene Symbol: Olfr1308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207151.1 | NP_997034.1 | 1 | 809 | ||
GenBank mRNA | BC108948.2 | - | 1 | 807 | ||
BC108949.2 | - | 1 | 808 | |||
BC115847.1 | - | 1 | 808 | |||
BC115949.1 | - | 1 | 808 | 40043972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
