siRNA Details
Gene Symbol: Olfr1337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146309.3 | NP_666421.2 | 1 | 405 | ||
GenBank mRNA | BC127989.1 | - | 1 | 401 | 40135644 | |
BC152318.1 | - | 1 | 407 |
Gene Symbol: Olfr1330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146334.2 | NP_666446.1 | 1 | 402 | ||
GenBank mRNA | BC108943.1 | - | 1 | 402 | 40042936 | |
BC108944.1 | - | 1 | 397 |
Gene Symbol: Olfr1333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207157.2 | NP_997040.1 | 1 | 396 | ||
GenBank mRNA | BC128000.1 | - | 1 | 400 | 40135841 |
Gene Symbol: Olfr1338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207152.2 | NP_997035.2 | 1 | 396 | ||
GenBank mRNA | BC130245.1 | - | 1 | 400 | 40142324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
