siRNA Details
Gene Symbol: Olfr114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146287.1 | NP_666399.1 | 1 | 813 | ||
GenBank mRNA | BC132209.1 | - | 1 | 843 | 40130486 | |
BC132573.1 | - | 1 | 843 | 40130850 |
Gene Symbol: Olfr115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011753.2 | NP_001011753.2 | 1 | 837 | ||
GenBank mRNA | BC139226.1 | - | 1 | 837 | 8862248 | |
BC139227.1 | - | 1 | 837 | 8862249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
