siRNA Details
Gene Symbol: MAPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002752.4 | NP_002743.3 | 8 | 1126 | ||
NM_139068.2 | NP_620707.1 | 8 | 1126 | |||
NM_139069.2 | NP_620708.1 | 8 | 1126 | |||
NM_139070.2 | NP_620709.1 | 8 | 1126 | |||
XM_005265940.4 | XP_005265997.1 | 9 | 1259 | |||
XM_006714891.2 | XP_006714954.1 | 9 | 1184 | |||
XM_017009638.1 | XP_016865127.1 | 9 | 1095 | |||
XM_017009639.1 | XP_016865128.1 | 10 | 1352 | |||
XM_017009640.1 | XP_016865129.1 | 9 | 1243 | |||
XM_017009641.1 | XP_016865130.1 | 9 | 1193 | |||
XM_017009642.1 | XP_016865131.1 | 8 | 1022 | |||
GenBank mRNA | AB451302.1 | - | 7 | 855 | ||
AB451355.1 | - | 7 | 855 | |||
AB451433.1 | - | 7 | 855 | |||
AK289638.1 | - | 8 | 1022 | |||
BC032539.1 | - | 7 | 916 | 5528624 | ||
CR536580.1 | - | 7 | 855 | |||
HM036707.1 | - | 7 | 875 | |||
HM036708.1 | - | 7 | 875 | |||
HM036709.1 | - | 8 | 1116 | |||
L31951.1 | - | 7 | 913 | |||
U09759.1 | - | 7 | 1006 | |||
U34821.1 | - | 7 | 899 | |||
U35002.1 | - | 7 | 877 | |||
U35003.1 | - | 7 | 889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
