siRNA Details
Gene Symbol: Olfr1329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011870.2 | NP_001011870.2 | 1 | 578 |
Gene Symbol: Olfr1337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146309.3 | NP_666421.2 | 1 | 587 | ||
GenBank mRNA | BC127989.1 | - | 1 | 583 | 40135644 | |
BC152318.1 | - | 1 | 589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
