siRNA Details
Gene Symbol: Olfr725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146317.2 | NP_666429.2 | 1 | 694 | ||
GenBank mRNA | BC125411.1 | - | 1 | 694 | 40129926 | |
BC125417.1 | - | 1 | 694 | 40129932 |
Gene Symbol: Olfr726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146316.2 | NP_666428.2 | 1 | 595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
