siRNA Details
Gene Symbol: Olfr187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146322.2 | NP_666434.2 | 2 | 357 | ||
GenBank mRNA | BC119304.1 | - | 1 | 245 | 8734053 | |
BC119306.1 | - | 1 | 248 | 8734055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
