siRNA Details
Gene Symbol: Olfr1128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146349.2 | NP_666461.2 | 1 | 406 | ||
GenBank mRNA | BC104064.2 | - | 1 | 405 | ||
BC104065.1 | - | 1 | 406 | 40041018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
