siRNA Details
Gene Symbol: Olfr677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146358.1 | NP_666470.1 | 1 | 654 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001001032.
Gene Ontology Categories:
