siRNA Details
Gene Symbol: Olfr1094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146365.2 | NP_666477.1 | 1 | 358 | ||
GenBank mRNA | BC120867.1 | - | 1 | 358 | 40129790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
