siRNA Details
Gene Symbol: Olfr228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146405.2 | NP_666517.2 | 1 | 326 |
Gene Symbol: Olfr1076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146406.2 | NP_666518.2 | 1 | 326 |

More Information
Other Species Targeted

Gene Ontology Categories:
