siRNA Details
Gene Symbol: Olfr291 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146415.2 | NP_666527.1 | 3 | 731 | ||
GenBank mRNA | BC100693.1 | - | 1 | 525 | ||
BC100694.1 | - | 1 | 524 | |||
BC100695.1 | - | 1 | 525 | |||
BC114974.1 | - | 1 | 525 | 40043317 | ||
BC115740.1 | - | 1 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
