siRNA Details
Gene Symbol: Olfr742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146430.2 | NP_666641.2 | 1 | 719 | ||
GenBank mRNA | BC132583.1 | - | 1 | 719 | 40130860 |
Gene Symbol: Olfr741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207133.2 | NP_997016.2 | 3 | 842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
