siRNA Details
Gene Symbol: Olfr996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146437.2 | NP_666648.2 | 1 | 804 | ||
GenBank mRNA | BC120782.1 | - | 1 | 804 | 40129705 | |
BC120784.1 | - | 1 | 804 | 40129707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
