siRNA Details
Gene Symbol: Olfr458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146444.1 | NP_666655.1 | 1 | 454 | ||
GenBank mRNA | BC106796.1 | - | 1 | 454 | ||
BC106797.1 | - | 1 | 454 | |||
BC119415.1 | - | 1 | 454 | |||
BC119416.1 | - | 1 | 454 | |||
BC127219.1 | - | 1 | 449 | |||
BC127220.1 | - | 1 | 454 |
Gene Symbol: Olfr456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011528.2 | NP_001011528.2 | 1 | 474 | ||
GenBank mRNA | BC139108.1 | - | 1 | 474 | 8862130 | |
BC139109.1 | - | 1 | 474 | 8862131 |
Gene Symbol: Olfr457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146987.1 | NP_667198.1 | 1 | 454 | ||
GenBank mRNA | BC119371.1 | - | 1 | 478 | 8734120 | |
BC119373.1 | - | 1 | 478 | 8734122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
