siRNA Details
Gene Symbol: Olfr215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146446.1 | NP_666657.1 | 1 | 661 | ||
GenBank mRNA | BC125561.1 | - | 1 | 748 | 40130076 | |
BC125563.1 | - | 1 | 748 | 40130078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
