siRNA Details
Gene Symbol: Olfr1215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146459.2 | NP_666670.2 | 1 | 80 | ||
GenBank mRNA | BC127970.1 | - | 1 | 80 | 40135109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
