siRNA Details
Gene Symbol: Olfr1196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146464.2 | NP_666675.2 | 1 | 794 |
Gene Symbol: Olfr1197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005225.1 | NP_001005225.1 | 1 | 794 |
Gene Symbol: Olfr1200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005227.2 | NP_001005227.2 | 1 | 794 |
