siRNA Details
Gene Symbol: Olfr1388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146467.1 | NP_666678.1 | 1 | 785 | ||
GenBank mRNA | BC104274.1 | - | 1 | 784 | ||
BC104275.1 | - | 1 | 785 | 40041223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
