siRNA Details
Gene Symbol: Olfr1391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146468.1 | NP_666679.1 | 1 | 763 | ||
GenBank mRNA | BC127961.1 | - | 1 | 767 | 40134837 | |
BC148323.1 | - | 1 | 767 | 40134755 | ||
BC150143.1 | - | 1 | 767 | 40134757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
