siRNA Details
Gene Symbol: Olfr91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182714.2 | NP_874373.2 | 1 | 760 | ||
GenBank mRNA | BC108941.1 | - | 1 | 758 | ||
BC108942.1 | - | 1 | 755 | |||
BC119413.1 | - | 1 | 760 | 40042089 | ||
BC119414.1 | - | 1 | 760 | 40042091 |

More Information
Other Species Targeted

Gene Ontology Categories:
