siRNA Details
Gene Symbol: Olfr899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146479.1 | NP_666690.1 | 1 | 519 | ||
GenBank mRNA | BC104124.1 | - | 1 | 519 | 40044544 | |
BC104125.1 | - | 1 | 519 | 40044545 |
Gene Symbol: Olfr250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207552.2 | NP_997435.2 | 1 | 519 |
