siRNA Details
Gene Symbol: Olfr97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146512.1 | NP_666723.1 | 1 | 884 | ||
GenBank mRNA | BC134363.1 | - | 888 | 40134689 |
Gene Symbol: Olfr98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146510.1 | NP_666721.1 | 1 | 881 | ||
GenBank mRNA | BC119060.1 | - | 1 | 968 | 8733809 | |
BC120744.1 | - | 1 | 968 | 40129667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
