siRNA Details
Gene Symbol: Olfr523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146518.1 | NP_666729.1 | 1 | 880 | ||
GenBank mRNA | BC119240.1 | - | 1 | 923 | 8733989 | |
BC120529.1 | - | 1 | 923 | 40129452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
