siRNA Details
Gene Symbol: Olfr855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146524.2 | NP_666735.2 | 1 | 472 | ||
GenBank mRNA | BC137820.1 | - | 1 | 472 | 8860839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
