siRNA Details
Gene Symbol: PROP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006261.4 | NP_006252.3 | 1 | 376 | ||
GenBank mRNA | AF076215.1 | - | 1 | 376 | ||
BC069076.1 | - | 1 | 67 | 7216890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
