siRNA Details
Gene Symbol: Olfr866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146558.2 | NP_666769.1 | 1 | 201 | ||
GenBank mRNA | BC119369.1 | - | 1 | 201 | 8734118 | |
BC137762.1 | - | 1 | 201 | 8860781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
