siRNA Details
Gene Symbol: Olfr1015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146571.2 | NP_666782.2 | 1 | 154 | ||
GenBank mRNA | BC119114.1 | - | 1 | 154 | 8733863 | |
BC119142.1 | - | 1 | 154 | 8733891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
