siRNA Details
Gene Symbol: Olfr360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146622.1 | NP_666833.1 | 1 | 216 | ||
GenBank mRNA | BC110546.2 | - | 1 | 214 | ||
BC125238.1 | - | 1 | 214 | |||
BC125239.1 | - | 1 | 215 | |||
BC126918.1 | - | 1 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
