siRNA Details
Gene Symbol: Olfr1140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146642.2 | NP_666853.2 | 1 | 499 | ||
GenBank mRNA | BC120608.1 | - | 1 | 499 | 40129531 | |
BC120610.1 | - | 1 | 499 | 40129533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
