siRNA Details
Gene Symbol: Olfr418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146651.2 | NP_666862.2 | 1 | 175 | ||
GenBank mRNA | BC119223.1 | - | 1 | 175 | 8733972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
