siRNA Details
Gene Symbol: Olfr825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146677.1 | NP_666888.1 | 1 | 44 | ||
GenBank mRNA | BC119054.1 | - | 1 | 142 | 8733803 | |
BC119056.1 | - | 1 | 142 | 8733805 |
Gene Symbol: Olfr826 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146676.1 | NP_666887.1 | 1 | 44 |
