siRNA Details
Gene Symbol: Olfr235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146686.2 | NP_666897.2 | 1 | 560 | ||
GenBank mRNA | BC132238.1 | - | 1 | 630 | 40130515 | |
BC132388.1 | - | 1 | 630 | 40130665 |
Gene Symbol: Olfr1431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146414.1 | NP_666526.1 | 1 | 560 | ||
GenBank mRNA | BC133710.1 | - | 1 | 564 | 40142241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
