siRNA Details
Gene Symbol: Olfr411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146709.2 | NP_666920.2 | 1 | 354 | ||
GenBank mRNA | BC138439.1 | - | 1 | 354 | 8861459 | |
BC138440.1 | - | 1 | 354 | 8861460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
