siRNA Details
Gene Symbol: Olfr433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146717.2 | NP_666928.2 | 1 | 869 | ||
GenBank mRNA | AK154347.1 | - | 1685 | |||
BC119238.1 | - | 1 | 869 | 8733987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
