siRNA Details
Gene Symbol: Olfr424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146721.2 | NP_666932.2 | 1 | 850 | ||
GenBank mRNA | BC104096.1 | - | 1 | 850 | 40043031 | |
BC104097.1 | - | 1 | 850 | 40043033 |

More Information
Other Species Targeted

Gene Ontology Categories:
