siRNA Details
Gene Symbol: Olfr429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146722.2 | NP_666933.2 | 1 | 415 | ||
GenBank mRNA | BC127973.1 | - | 1 | 415 | 40135130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
