siRNA Details
Gene Symbol: Olfr483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146735.1 | NP_666946.1 | 1 | 491 | ||
GenBank mRNA | BC116910.1 | - | 1 | 523 | 40126229 | |
BC116912.1 | - | 1 | 523 | 40126231 |
