siRNA Details
Gene Symbol: Olfr502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146739.1 | NP_666950.1 | 1 | 638 | ||
GenBank mRNA | BC119235.1 | - | 1 | 657 | 8733984 | |
BC120678.1 | - | 1 | 657 | 40129601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
