siRNA Details
Gene Symbol: Olfr689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146750.1 | NP_666961.1 | 1 | 377 | ||
GenBank mRNA | BC119379.1 | - | 1 | 401 | 8734128 |
Gene Symbol: Olfr688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011533.2 | NP_001011533.2 | 1 | 380 |
