siRNA Details
Gene Symbol: Olfr551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146755.2 | NP_666966.1 | 1 | 833 | ||
GenBank mRNA | BC141014.1 | - | 1 | 834 | 9055626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
