siRNA Details
Gene Symbol: Olfr414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146761.2 | NP_666972.2 | 1 | 223 | ||
GenBank mRNA | BC106815.1 | - | 1 | 223 | 40043364 | |
BC106816.1 | - | 1 | 223 | 40043365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
