siRNA Details
Gene Symbol: Olfr922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146781.2 | NP_666992.1 | 1 | 327 | ||
GenBank mRNA | BC119109.1 | - | 1 | 327 | 8733858 | |
BC125623.1 | - | 1 | 327 | 40130138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
